Learn More
Invitrogen™ ANTS (8-Aminonaphthalene-1,3,6-Trisulfonic Acid, Disodium Salt)
Polyanionic dye often used in combination with the cationic quencher DPX for membrane fusion or permeability assays, including complement-mediated immune lysis
Brand: Invitrogen™ A350
Code : 50
Additional Details : Weight : 0.00525kg
Description
- The unusually high Stokes shift of ANTS in water (>150nm) separates this from much of the autofluorescence of biological samples
- ANTS has also been employed as a neuronal tracer
Order Info
Shipping Condition: Room temperature
Specifications
Room Temperature | |
Cell Membranes & Lipids | |
C10H7NNa2O9S3 | |
Green |
Fluorescence Microscope | |
Fluorescent | |
Solid | |
Cell Tracer |
For Research Use Only. Not for use in diagnostic procedures.