Learn More
Invitrogen™ ATP Determination Kit, 200-1,000 assays
Based on luciferases absolute requirement for ATP in producing light
Brand: Invitrogen™ A22066
Code : 50
Additional Details : Weight : 0.05000kg
Description
Molecular Probes ATP Determination Kit offers a convenient bioluminescence assay for quantitative determination of ATP with recombinant firefly luciferase and its substrate D-luciferin. The assay is based on luciferase's absolute requirement for ATP in producing light (emission maximum approx.560nm at pH 7.8). This assay is extremely sensitive; most luminometers can detect as little as 0.1 picomole of pre-existing ATP, or ATP as it is being formed in kinetic systems. This sensitivity has led to numerous applications for detecting ATP production in various enzymatic reactions, including ATPase and NADPH oxidase, as well as for detecting low-level bacterial contamination in samples such as blood, milk, urine, soil and sludge. The luciferin–luciferase bioluminescence assay has also been used successfully to study the effects of antibiotics on bacterial populations, to determine cell proliferation and cytotoxicity in both bacterial and mammalian cells, and to distinguish cytostatic versus cytocidal potential of anticancer drugs on malignant cell growth.
- Luminescence is monitored at approx. 560nm using a luminometer
- Can detect as little as 0.1 picomole of pre-existing ATP, or ATP as it is being formed in kinetic systems
- Luciferase and luciferin are packaged separately, which enables researchers to optimize conditions for their samples and instruments
- Includes sufficient reagents to perform 200 ATP assays using 500μL sample volumes or 1000 ATP assays using 100μL sample volumes
Apoptosis, Biochemical Toxicology Assays, Cell Analysis, Cell Culture, Cell Cycle, Cell Metabolism, Cell Proliferation, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Drug Discovery and Development, High-Content Screening (HCS), Multiparametric Apoptosis Analysis, Stem Cell Analysis, Stem Cell Labeling and Detection, Stem Cell Research, Target-Based ADME/Tox Assays
Order Info
Shipping Condition: Wet ice
Specifications
Bioluminescent, Chemiluminescent | |
Luminometer (Microplate), Luminometer (Tube-Based), Microplate Reader | |
Store in freezer -5°C to -30°C and protect from light. | |
D-Luciferin |
Tube(s), 96-well plate | |
ATP Determination Kit | |
Wet Ice | |
Luciferin |
For Research Use Only. Not for use in diagnostic procedures.