Learn More
Invitrogen™ BackDrop™ Background Suppressor for Microplate Assays
Designed to effectively suppress background fluorescence in live cell assays
Brand: Invitrogen™ B10512
252.32 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
BackDrop Background Suppressor is a novel signal enhancer reagent that helps improve signal-to-noise ratios when used with green-emitting fluorescent dyes or proteins.
- Can be added to complete media
- Improves sensitivity of green-emitting fluorescent dyes or proteins
- Validated with many cellular assays, including ion indicator and membrane potential assays
- Minimal cell toxicity—demonstrated to not cause osmotic shock to neuronal cells
The sensitivity of fluorescent assays can be compromised by autofluorescence or background fluorescent signals. These background signals result from cellular proteins or growth media components and contribute to a decrease in the signal-to-noise ratio and reduced assay sensitivity. Addition of BackDrop Background Suppressor significantly reduce background signal, helping improve experimental results. BackDrop Background Suppressor can be added to growth media and has been formulated to not contribute to cell toxicity. It has been successfully used with many different cell types and fluorescent reagents (including ion indicators) to efficiently suppress background fluorescence.
Cell Analysis, Cellular Imaging, Immunocytochemistry (ICC)
Specifications
One 50 mL bottle | |
Store at 2-8°C. DO NOT FREEZE. Protect material from long-term exposure to light. | |
BackDrop™ | |
Background Suppressor for live cell assays |
50 mL | |
Liquid | |
Microplate Assays |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.