missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BacMam GFP Transduction Control (BacMam 2.0)

Important part of the enhanced BacMam technology

Brand:  Invitrogen™ B10383

239.58 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10754639

  • £363.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: BacMam GFP transduction control contains 1mL of virus at a concentration of approximately 1 x 108 viral particles/mL. Approximate fluorescence excitation/emission maxima for emGFP: 485/520 in nm.
This item is not returnable. View return policy

Description

Description

The BacMam 2.0 Transduction Control lets you experience the power and convenience of enhanced BacMam technology, with the convenient readout of emerald GFP (emGFP); our Aequorea victoria emGFP is extremely bright and photostable and this non-targeted form is ideal for testing transduction. Check out how well BacMam 2.0 works with GFP in your model before advancing to the more specialized reagents. The BacMam technology is based on an insect virus (baculovirus) to help efficiently deliver and express genes in mammalian cells. The baculovirus has been modified to include an expression cassette for transgene expression in mammalian cells. BacMam 2.0 incorporate elements that help greatly enhance transduction efficiency and expression levels: a pseudotyped capsid protein for more efficient cell entry and genetic elements (enhanced CMV promoter and Woodchuck Post-transcriptional Regulatory Element), that boost expression levels. Baculoviruses do not replicate in mammalian cells and thus have an excellent safety profile and lack cytopathic effects on cells. Cell types previously not compatible with the technology (neuronal cells, T-cells), or only poorly transduced (some stem cells, CHO) are now transduced quantitatively in a simple, one step process.

  • Efficient: highly efficient (>90%) transduction of a wide range of mammalian cell lines, including primary cells, stem cells and neuronal cells
  • Fast and convenient: Simply add the BacMam 2.0 together with cells to your cover slip or microtiter plate, incubate overnight and assay or store frozen, assay-ready cells for later use
  • Safe: non-replicating in mammalian cells, lack of observable cytopathic effect and biosafety level (BSL) 1 handling
  • Enables Flexiblity: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies and tightly control expression levels by just varying the dose

Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Fluorescent Protein Assays (GFP)

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Green
485⁄520
Fluorescence Microscope, Fluorometer, HTS Reader, High Content Instrument, Flow Cytometer
Fluorescence
Liquid
Wet Ice
Fluorescent Dye
Cytoplasm, Nucleus
The BacMam GFP ttransduction control contains 1 ml of virus at a concentration of approximately 1 × 10^8 viral particles⁄ml.
Approximate fluorescence excitation⁄emission maxima for emGFP: 485⁄520 in nm.
The BacMam transduction control should be stored at 4°C, protected from light. When stored as directed, it is stable for 6 months. DO NOT FREEZE.
GFP (EmGFP)
1 vial
Visible
CMV
Fluorescence Intensity
BacMam GFP Transduction Control
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.