missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAPTA, Tetrasodium Salt, cell impermeant

A cell-impermeant chelator, which is a highly selective for Ca2+ over Mg2+

Brand:  Invitrogen™ B1214

Product Code. 10114232

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Can be used to control the level of extracellular Ca2+
  • BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
BAPTA Tetrasodium Salt
Chelator (Non-Fluorescent)
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2306583 Certificate TypeCertificate of Analysis Date23/12/2020 Catalog NumberB1214
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ BAPTA, Tetrasodium Salt, cell impermeant > 1g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.