missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BCECF, AM (2',7'-Bis-(2-Carboxyethyl)-5-(and-6)-Carboxyfluorescein, Acetoxymethyl Ester)

For measuring changes in the cytosolic pH of most cells

£224.00 - £300.00

Specifications

Content And Storage Store in freezer -5°C to -30°C.
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Product Type BCECF AM pH Indicator
Dye Type pH Indicator
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11524147
View Documents
Invitrogen™
B1170
20 x 50 μg
£300.00
1000µg
Please to purchase this item. Need a web account? Register with us today!
10646693
View Documents Promotion Details
Invitrogen™
B1150
1mg
£224.00
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • With pKa of approximately 6.98, cell-permeant, dual-excitation ratiometric pH indicator, AM is ideal for measuring changes in cytosolic pH of most cells
  • Intracellular pH measurements with BCECF are made by determining pH-dependent ratio of emission intensity (detected at 535nm) when dye is excited at approximately 490nm versus emission intensity when excited at its isobestic point of approximately 440nm

Cell Adhesion, Cell Analysis, Cell Metabolism, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cell-Based Ion Channel Assays, Drug Discovery and Development, Ion Channel Biology, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation, pH Detection

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Store in freezer -5°C to -30°C.
Microplate Reader
pH Indicator
Fluorescence
BCECF AM pH Indicator
Room Temperature
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.