Learn More
Invitrogen™ Bis-ANS (4,4'-Dianilino-1,1'-Binaphthyl-5,5'-Disulfonic Acid, Dipotassium Salt)
Binds with affinity that is often orders-of-magnitude higher than 1-8-ANS as probe for nonpolar cavities in proteins
Brand: Invitrogen™ B153
Code : 50
Additional Details : Weight : 0.00525kg
Description
Anilinonaphthalene sulfonates (ANS) are essentially nonfluorescent in water, only becoming appreciably fluorescent when bound to membranes. This property makes it a sensitive indicator of protein folding, conformational changes and other processes that modify the exposure of the probe to water.
Cell Analysis, Cell Structure, Cellular Imaging, Cytoskeleton, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Room Temperature | |
Molecular Probes™ | |
C32H22K2N2O6S2 | |
Protein Folding Indicator |
Cell Membranes & Lipids | |
Other Label(s) or Dye(s) | |
Solid |
For Research Use Only. Not for use in diagnostic procedures.