Learn More
Invitrogen™ Bis-BODIPY™ FL C11-PC) (1,2-Bis-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Undecanoyl)-sn-Glycero-3-Phosphocholine)
Dye-labeled phospholipids used to monitor phospholipase A (PLA) activity
Brand: Invitrogen™ B7701
220.67 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Description
Molecular Probes phospholipase substrates are dye labeled phospholipids used to monitor phospholipase A (PLA) activity in purified enzyme preparations, cell lysates, and living cells. Two structural variations of these substrates are available, which confer specificity for PLA1 and PLA2. Fluorescently labeled versions of the cleavage product generated from these substrates are also available for use as standards for evaluating percentage substrate conversion in enzymatic reactions.
- Label (Ex/Em): Bis-BODIPY FL C11-PC (approx. 488/530nm)
- Specificity: PLA1 or PLA2
- Cleavage product: D3862
- Lyophilized product can be dissolved in ethanol for use
- Fluorescence is measured either in microplate assays or using flow cytometry
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature
Specifications
Phospholipids | |
100 μg | |
Solid | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.