Learn More
Invitrogen™ BODIPY™ 558/568 C12 (4,4-Difluoro-5-(2-Thienyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoic Acid)
The orange-red fluorescent fatty acid, BODIPY™ 558/568 C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.
Brand: Invitrogen™ D3835
Code : 50
Additional Details : Weight : 0.00525kg
Description
The orange-red fluorescent fatty acid, BODIPY™ 558/568 C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY™ dyes are relatively environment insensitive and is fluorescent in both aqueous and lipid environments.
We recommend dissolving in high-quality anhydrous DMSO to attain a stock concentration up to 1 mM.
Order Info
Shipping Condition: Room Temperature
Specifications
Fatty Acids | |
Solid | |
559/568 nm |
1 mg | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.