Learn More
Invitrogen™ BODIPY™ FL C12-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoyl) Sphingosyl Phosphocholine)
Green-fluorescent sphingomyelin
Brand: Invitrogen™ D7711
Code : 50
Additional Details : Weight : 0.00450kg
Description
BODIPY FL C12-sphingomyelin can be used to study sphingolipid transport and signaling pathway in cells.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Sphingolipids | |
250 μL | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.