Learn More
Invitrogen™ BODIPY™ FL C12 (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoic Acid)
The green fluorescent fatty acid, BODIPY™ FL C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.
Brand: Invitrogen™ D3822
Code : 50
Additional Details : Weight : 0.00525kg
Description
The green fluorescent fatty acid, BODIPY™ FL C12 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY™ dyes are relatively environment insensitive and is fluorescent in both aqueous and lipid environments.
Dissolve in high-quality anhydrous DMSO for stock concentrations of 1 to 10 mM.
Order Info
Shipping Condition: Room Temperature
Specifications
Fatty Acids | |
418.33 | |
BODIPY |
1 mg | |
Solid |
For Research Use Only. Not for use in diagnostic procedures.