Learn More
Invitrogen™ BODIPY™ FL C5 (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoic Acid)
Fluorescent in both aqueous and lipid environments
Brand: Invitrogen™ D3834
Code : 50
Additional Details : Weight : 0.00525kg
Description
The green fluorescent fatty acid, BODIPY FL C5 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY dyes are relatively environment insensitive.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Fatty Acids | |
1 mg | |
Solid | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.