Learn More
Invitrogen™ BODIPY™ FL C5-Ceramide complexed to BSA
Can be used in sphingolipid transport and metabolism mechanisms
Brand: Invitrogen™ B22650
Code : 50
Additional Details : Weight : 0.00454kg
Description
Complexing fluorescent lipids with bovine serum albumin (BSA) facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe - the BSA-complexed probe can be directly dissolved in water.
Cell Analysis, Cell Structure, Golgi
Order Info
Shipping Condition: Room Temperature
Specifications
Sphingolipids | |
5 mg | |
Solid | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.