Learn More
Invitrogen™ BODIPY™ FL C5-Ganglioside GM1
Useful in cellular imaging
Brand:  Invitrogen™ B13950
262.96 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 11504157
  • £346.00 / 25µg
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Colocalization of fluorescent cholera toxin B conjugates and the green-fluorescent BODIPY FL C5-ganglioside GM1 observed by fluorescence microscopy provides a direct indication of the association of these molecules in lipid rafts.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sphingolipids
25 μg
Solid
BODIPY