Learn More
Invitrogen™ BODIPY™ FL C5-Lactosylceramide complexed to BSA
The green-fluorescent BODIPY™ FL C5 lactosylceramide complexed to BSA can be used in sphingolipid transport and metabolism mechanisms.
Brand: Invitrogen™ B34402
285.00 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
The green-fluorescent BODIPY™ FL C5 lactosylceramide complexed to BSA can be used in sphingolipid transport and metabolism mechanisms. Complexing fluorescent lipids with bovine serum albuin (BSA) facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe—the BSA-complexed probe can be directly dissolved in water.
Consult user Manual for solubility instructions.
Order Info
Shipping Condition: Room Temperature
Specifications
Sphingolipids | |
Solid | |
505/511 nm |
1 mg | |
BODIPY |