Learn More
Invitrogen™ BODIPY™ FL C5-Sphingomyelin (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)Sphingosyl Phosphocholine)
Useful in cellular imaging
Brand: Invitrogen™ D3522
Code : 50
Additional Details : Weight : 0.00525kg
Description
BODIPY FL C5 sphingomyelin can be used in sphingolipid transport and metabolism studies.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Sphingolipids | |
250 μg | |
Solid | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.