Learn More
Invitrogen™ BODIPY™ FL DHPE (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionyl)-1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)
Can form excimers at high concentrations
Brand: Invitrogen™ D3800
157.34 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
BODIPY DHPE has the green-fluorescent dye attached to the head group of the phospholipid. Unlike fluorophores like pyrene, DPH and NBD, BODIPY FL is relatively environment insensitive and is fluorescent in both aqueous and lipid environments.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Phospholipids | |
100 μg | |
Solid | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.