missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BODIPY™ FL Prazosin
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Prazosin is a high-affinity antagonist for the a1-adrenergic receptor. The green-fluorescent BODIPY FL prazosin derivative can be used to localize and detect the a1-adrenergic receptor.
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Glutathione Detection, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Nitro-Oxidative Stress
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Quantity | 100 μg |
| Shipping Condition | Room Temperature |
| Product Line | BODIPY |
| Label Type | BODIPY Dyes |
| Molecular Formula | C28H32BF2N7O3 |
| Product Type | Antagonist |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction