Learn More
Invitrogen™ BODIPY™ TR Ceramide complexed to BSA
Can be used in sphingolipid transport and metabolism mechanisms
Brand:  Invitrogen™ B34400
304.00 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 11514187
  • £400.00 / 5mg
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
The red-fluorescent BODIPY TR C5 ceramide complexed to bovine serum albumin. Complexing fluorescent lipids with BSA facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe; the BSA-complexed probe can be directly dissolved in water.
Cell Analysis, Cell Structure, Golgi
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sphingolipids
5 mg
Solid
BODIPY