Learn More
Invitrogen™ BrdU (5-Bromo-2'-Deoxyuridine)
Excellent marker of cell cycle and proliferation
Brand: Invitrogen™ B23151
111.30 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Description
Cells can naturally incorporate the thymidine analog BrdU into their cells during cell division, making this nucleoside analog an excellent marker of by cell cycle and proliferation. Analysis of incorporated BrdU can be with an anti-BrdU antibody.
Apoptosis, Cell Analysis, Cell Cycle, Cell Proliferation, Cell Viability, Proliferation and Function, DNA Fragmentation, Flow Cytometry, Flow Cytometry Apoptosis Assays, Flow Cytometry Cell Proliferation, Flow Cytometry of Cellular Processes
Order Info
Shipping Condition: Room temperature

Specifications
BrdU | |
Room Temperature | |
100 mg |
Store in freezer -5°C to -30°C and protect from light. | |
BrdU |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.