Learn More
Invitrogen™ CellTracker™ Red CMTPX Dye
A fluorescent dye well suited for monitoring cell movement or location
Brand: Invitrogen™ C34552
336.68 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price
Description
- After loading cells, dye is well retained, allowing for multigenerational tracking of cellular movements
- Deep red excitation/emission spectra (577/602nm maxima) are optimal for multiplexing with green fluorescent dyes and proteins
- Easy to use: remove medium, add dye, incubate 30 minutes, and image cells
- Fluorescent signal retention of >72 hours (typically three to six generations)
- Low cytotoxicity: does not affect viability or proliferation
- Designed to freely pass through cell membranes into cells, where it is transformed into a cell-impermeant, fluorescent dye
- Dye is transferred to daughter but not adjacent cells in population
- Stable, nontoxic at working concentrations, well retained in cells, and brightly fluorescent at physiological pH
Cell Adhesion, Cell Analysis, Cell Proliferation, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Cellular Toxicology Assays, Chemotaxis and Cell Migration, Drug Discovery and Development, General Cell Tracing, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Target-Based ADME/Tox Assays
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C and protect from light. | |
CellTracker™ | |
Room Temperature | |
Dye |
577⁄602 | |
Cell Tracker Compounds, Cell Labeling Reagents | |
Other Labels or Dyes |
For Research Use Only. Not for use in diagnostic procedures.