Learn More
Invitrogen™ CholEsteryl 4,4-Difluoro-5-(4-Methoxyphenyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-Undecanoate (CholEsteryl BODIPY™ 542/563 C11)
Orange-red fluorescent cholesteryl ester can be used as tracer of cholesterol transport and rand as general nonexchangeable membrane marker
Brand: Invitrogen™ C12680
Code : 50
Additional Details : Weight : 0.00525kg
Description
Cholesteryl esters consist of a fatty acid esterified to the 3β-hydroxyl group of cholesterol. These very nonpolar species are the predominant lipid components of atherosclerosis plaque and low- and high-density lipoporotein (LDL and HDL) cores.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Cholesterol Analogs | |
1 mg | |
Solid | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.