Learn More
Invitrogen™ CholEsteryl 4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoate (CholEsteryl BODIPY™ FL C12)
Can be used as general nonexchangeable membrane marker
Brand: Invitrogen™ C3927MP
Code : 50
Additional Details : Weight : 0.05250kg
Description
Cholesteryl esters consist of a fatty acid esterified to the 3β-hydroxyl group of cholesterol. These very nonpolar species are the predominant lipid components of atherosclerosis plaque and low- and high-density lipoporotein (LDL and HDL) cores. The green fluorescent BODIPY™ FL C12 cholesteryl ester can be used as a tracer of cholesterol transport and receptor-mediated endocytosis of lipoproteins by fluorescence microscopy.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Cholesterol Analogs | |
1 mg | |
Solid | |
BODIPY |
For Research Use Only. Not for use in diagnostic procedures.