missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EU (5-Ethynyl Uridine)

Product Code. 10726865
Click to view available options
Quantity:
5 mg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10726865

Brand: Invitrogen™ E10345

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Aids in detection of global RNA transcription

Detect global RNA transcription temporally and spatially in both in-vitro and in-vivo with the modified nucleotide, ethynyl uridine. Approach uses powerful click chemistry detection, a simple and robust two-step technique. In step one, the alkyne-containing biomolecule is fed to cells or animals and actively incorporated into RNA. Detection utilizes the chemoselective ligation or 'click' reaction between an azide and an alkyne where the modified protein is detected with a corresponding azide-containing dye or hapten using the Click-iT Cell Reaction Buffer Kit. Cells can be analyzed by fluorescence microscopy, flow cytometry or high-content imaging and analysis (HCS).

Cell Analysis, Click Chemistries, Labeling Chemistry, Metabolic Labeling and Nascent Protein Synthesis with Click Chemistry

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type EU (5-Ethynyl Uridine)
Content And Storage Store at ≤-20°C
Labeling Method RNA Labeling
Shipping Condition Room Temperature
Label or Dye EU
Quantity 5 mg
Product Line Click-iT, Molecular Probes

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.