Learn More
Invitrogen™ Click-IT™ O-GlcNAc Enzymatic Labeling System
For in-vitro modification of O-GlcNAc modified proteins
Brand: Invitrogen™ C33368
238.01 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The Click-iT O-GlcNAc Enzymatic Labeling System provides a highly sensitive and efficient method for the in-vitro modification of O-GlcNAc modified proteins. Proteins are enzymatically labeled utilizing the permissive mutant β-1,4-galactosyltransferase (Gal-T1 (Y289L)) which transfers azido-modified galactose (GalNAz) from UDP-GalNAz to O-GlcNAc residues. Then, via the chemoselective ligation or click reaction between an azide and an alkyne, the azido-labeled glycoproteins can then be detected with a Click-iT Glycoprotein Detection kit for gels (TAMRA or Dapoxyl™ alkyne) or Western blots (biotin alkyne). Labeling and detection can be completed in less than 24 hours and the Click-iT glycoprotein products are compatible with LC-MS/MS and Multiplexed Proteomics™ technologies for in-depth analyses of this important post-translational modification.
- Less hazardous
Cell Analysis, Cell Structure, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Mitochondria, Mitochondria Related Diseases, Mitochondrial Pathways, Protein and Antibody Labeling, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet ice
Specifications
Less hazardous | |
1 Kit | |
Biotin-based, Fluorescence | |
Click-iT™ | |
Proteins |
O-GlcNAc Enzymatic Labeling System | |
Store in refrigerator at 2°C to 8°C | |
Wet Ice | |
Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647, Biotin, Oregon Green 488, TMR (Tetramethylrhodamine) |
For Research Use Only. Not for use in diagnostic procedures.