Learn More
Invitrogen™ Click-iT™ Protein Reaction Buffer Kit
Click two moieties together: azide or alkyne and the corresponding click detection reagent
Brand: Invitrogen™ C10276
240.50 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
The Click-iT Protein Buffer Kit provides researchers who have proteins labeled with an azide or alkyne and the corresponding click detection reagent with the means to click these two moieties together. Each kit includes sufficient material to perform 25 reactions for subsequent analysis by gel electrophoresis, western blot or mass spectrometry.
- Less hazardous
Cell Analysis, Click Chemistries, Labeling Chemistry, Metabolic Labeling and Nascent Protein Synthesis with Click Chemistry
Order Info
Shipping Condition: Room temperature
Specifications
Less hazardous | |
25 Reactions | |
Room Temperature |
Protein Reaction Buffer Kit | |
Store at room temperature. | |
Click-iT™, Molecular Probes™ |
Safety and Handling
- Click-iT Protein Reaction Buffer Kit
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Copper (II) sulfatepentahydrate
For Research Use Only. Not for use in diagnostic procedures.