Learn More
Invitrogen™ CoverWell™ Incubation Chamber Gasket
Designed for immunocytochemistry and in situ hybridization
Brand: Invitrogen™ C18150
Code : 50
Additional Details : Weight : 0.00450kg
Description
These heat resistant, autoclavable and nuclease free chamber gaskets are designed for use with incubation chambers. The chambers improve staining uniformity and sensitivity by enclosing a large sample area while minimizing the reagent volume required. The watertight seal holds reactants in place and prevents evaporation even during prolonged heating and is easily removed for multiple-step procedures.
Applications:
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunofluorescence Staining and Detection, Immunohistochemistry (IHC)
Specifications
Incubation Chamber Gasket | |
0.2 mm | |
0.2 mm | |
40 mm x 22 mm |
Red | |
Store at room temperature. | |
Room Temperature | |
CoverWell |
For Research Use Only. Not for use in diagnostic procedures.