Learn More
Invitrogen™ CoverWell™ Perfusion Chamber Gasket, 8 chambers, 9 mm diameter, 0.5 mm deep
Designed to facilitate live-cell imaging and manipulation
Brand: Invitrogen™ C18139
150.33 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
- CoverWell perfusion chamber gaskets form watertight “press-to-seal” chambers with dual-access ports so reagents can be quickly added and removed
- Available in single- or multi-well configurations, these gaskets allow multiple experiments to be performed on single microscope slide or coverslip
- Heat-resistant gaskets can be sterilized and used for direct culturing of cells and tissues
Applications:
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Immunohistochemistry (IHC)
Specifications
Perfusion Chamber Gasket | |
0.5 mm | |
0.5 mm | |
Room Temperature | |
Sterile |
Red | |
Store at room temperature. | |
9 mm | |
CoverWell |
For Research Use Only. Not for use in diagnostic procedures.