Learn More
Invitrogen™ CultureWell™ Chambered Coverglass for Cell Culture, 16 wells per coverglass
Ideal for cell culture and fluorescence imaging applications
Brand: Invitrogen™ C37000
209.95 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
The CultureWell removable chambered coverglass provided sterile and ready to use—contains 16 wells that can each hold up to 250μL, allowing cells to be cultured in a number of different conditions on a single slide. When the cells are ready to be imaged, coverglass removal is made easy by the use of a simple tool (Cat. No. C37010, purchased separately) that separates the parts without the need for excessive force, eliminating the risk of coverglass breakage. Frosted microscope slides are also provided for mounting. A sample size containing two chambered coverglasses and the removal tool is also available (Cat. No. C37005)
Cell Analysis, Cell Culture, Cellular Imaging, IHC Sample Preparation, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC), Mammalian Cell Culture
Specifications
Coverglass | |
0.4 cm2 | |
250 μL | |
Room Temperature |
16 | |
Cell Culture | |
Store at room temperature. | |
Sterile |
For Research Use Only. Not for use in diagnostic procedures.