Learn More
Invitrogen™ D-Luciferin, Potassium Salt
Needed for luciferin/luciferase light-producing reaction
Brand: Invitrogen™ L2916
Code : 50
Additional Details : Weight : 0.00525kg
Description
The ATP-dependent oxidation of luciferin by luciferase produces light, and this system is employed as a reporter in plants, bacteria, and mammalian cells. Because chemiluminescent techniques are virtually background-free, this reporter gene system is ideal for detecting low-level gene expression.
- Exhibits physical properties identical to those of natural luciferin
- Chemiluminescent after oxidation by luciferase
- Luciferin potassium salt is readily soluble in aqueous buffers
Find More Fluorogenic and Photoactivatable Probes
A number of fluorogenic substrates, photoactivatable probes, and caged probes are available.
Cell Analysis, Enzyme and Protein Activity Assays, Luciferase Assays, Proteins, Expression, Isolation and Analysis, Reporter Assays
Order Info
Shipping Condition: Room temperature
Specifications
D-Luciferin | |
Store in freezer -5°C to -30°C and protect from light. | |
Room Temperature | |
Luciferase Substrate |
25 mg | |
Chemiluminescent | |
Chemical Substrate | |
Luciferin |
For Research Use Only. Not for use in diagnostic procedures.