missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dextran, Fluorescein, 10,000 MW, Anionic

Product Code. 11510226
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11510226

Brand: Invitrogen™ D1821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rigorous methods for removing as much unconjugated dye as practical, followed by an assay of dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

  • Succinimidyl coupling of dyes to dextran molecule, which, in most cases, results in a neutral or anionic dextran
  • Suitable for neuronal tracing (anterograde and retrograde) in live cells
  • Cell lineage tracing in live cells
  • Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
  • Investigating vascular permeability and blood-brain barrier integrity
  • Monitoring acidification (some dextran-dye conjugates are pH-sensitive)
  • Studying hydrodynamic properties of cytoplasmic matrix

  • The fluorescein dextrans are intrinsically anionic, whereas most of dextrans labeled with zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral
  • To produce more highly anionic dextrans, proprietary procedure adds negatively charged groups to the dextran carriers; these products are designated polyanionic dextrans

Cell Analysis, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, General Cell Tracing, Ionic Homeostasis and Signaling, Neuronal Tracing, pH Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Dextran
Quantity 25 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye Classic Dyes
Shipping Condition Room Temperature
Excitation/Emission 494/521 nm
Product Line Invitrogen

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.