missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DiFMUP (6,8-Difluoro-4-Methylumbelliferyl Phosphate)

Product Code. p-4921019
Click to view available options
Quantity:
5mg
10 x 10mg
Unit Size:
100mg
5mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11560346

Brand: Invitrogen™ D6567

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Continuously assay acid phosphatases at low pH

DiFMUP is a fluorinated 4-methylumbelliferyl phosphate (MUP) derivative that has a lower pKa than that of MUP, making DiFMUP an excellent substrate for continuously assaying acid phosphatases at low pH. The reaction product of DiFMUP has excitation/emission maxima of approx. 358/450nm.

Biochemical Kinase Assays, Biochemical Toxicology Assays, Cell Analysis, Cellular Imaging, Cellular Toxicology Assays, Drug Discovery and Development, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Kinase Biology, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation, Target-Based ADME/Tox Assays

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Powder
Quantity 5mg
Substrate Phosphatase Substrate
Substrate Properties Chemical Substrate
Target Enzyme Phosphatases
Excitation/Emission 358⁄455
For Use With (Equipment) Microplate Reader
Label or Dye DiFMU
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.