Learn More
Invitrogen™ Dihydroethidium (Hydroethidine)
Free radical sensor
Brand: Invitrogen™ D23107
Code : 50
Additional Details : Weight : 0.00525kg
Description
- Superoxide indicator dihydroethdium, also called hydroethidine, exhibits blue-fluorescence in cytosol until oxidized, where it intercalates within cell's DNA, staining its nucleus a bright fluorescent red
- Excitation Emission: 518/606nm
Cat. No. D23107 packaged 5mM solution stabilized in DMSO.
Apoptosis, Cell Analysis, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Free Radical Detection, Nitro-Oxidative Stress, Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing, Reactive Oxygen Species
Specifications
1 mL, 5 mM solution stabilized in DMSO | |
518/606 nm |
For Research Use Only. Not for use in diagnostic procedures.