Learn More
Invitrogen™ Dihydrorhodamine 123
Uncharged and nonfluorescent reactive oxygen species indicator
Brand: Invitrogen™ D632
Code : 50
Additional Details : Weight : 0.00525kg
Description
Dihydrorhodamine 123 can passively diffuse across membranes where it is oxidized to cationic rhodamine 123 which localizes in the mitochondria and exhibits green fluorescence.
Concentration: 5mM
Applications:
Apoptosis, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Free Radical Detection, Nitro-Oxidative Stress, Reactive Oxygen Species
Specifications
10 mg | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.