missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DMNPE-caged ATP (Adenosine 5'-Triphosphate, P3-(1-(4,5-Dimethoxy-2-Nitrophenyl)ethyl) Ester, Disodium Salt)

Flash photolysis of the caging group yields highly localized release of the free nucleotide

Brand:  Invitrogen™ A1049

Product Code. 11539046

This item is not returnable. View return policy

Description

Description

DMNPE-caged ATP is a nucleotide analog in which the terminal phosphate is esterified with a blocking group, rendering the molecule biologically inactive. Flash photolysis of the caging group with UV illumination at 355nm results in a rapid and highly localized release of the free nucleotide at the site of illumination. The DMNPE caging group absorbs light more efficiently than CNB and NPE caging groups at approx. 360nm, but photolysis rates and quantum yields are generally lower.

Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DMNPE-Caged ATP
Wet Ice
5 mg
Store in freezer -5°C to -30°C and protect from light.
DMNPE (P3-(1-(4,5-dimethoxy-2-nitrophenyl)ethyl) ester)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2413892 Certificate TypeCertificate of Analysis Date10/11/2021 Catalog NumberA1049
Lot Number2300984 Certificate TypeCertificate of Analysis Date28/05/2021 Catalog NumberA1049
Lot Number2161815 Certificate TypeCertificate of Analysis Date09/10/2019 Catalog NumberA1049
Lot Number1901206 Certificate TypeCertificate of Analysis Date02/08/2017 Catalog NumberA1049
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ DMNPE-caged ATP (Adenosine 5'-Triphosphate, P3-(1-(4,5-Dimethoxy-2-Nitrophenyl)ethyl) Ester, Disodium Salt) > 5mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.