Learn More
Invitrogen™ DPX (p-Xylene-Bis-Pyridinium Bromide)
Cationic quencher
Brand: Invitrogen™ X1525
Code : 50
Additional Details : Weight : 0.00525kg
Description
DPX (p-xylene-bis-pyridinium bromide) can be used in membrane fusion or permeability assays with the polyanionic, polar tracer dye ANTS (A350).
Cell Analysis, Cell Tracing & Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Room Temperature | |
C18H18Br2N2 | |
Solid | |
DPX Quencher |
For Research Use Only. Not for use in diagnostic procedures.