missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DQ™ Collagen, type I From Bovine Skin, Fluorescein Conjugate

The fluorogenic DQ™ collagen can be used to directly monitor collagenase activity.

Brand:  Invitrogen™ D12060

Code : 50

Additional Details : Weight : 0.00525kg

Product Code. 10562903

  • £384.00 / 1mg
In Stock
to see stock.
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

The fluorogenic DQ™ collagen can be used to directly monitor collagenase activity. DQ™ substrates are analogs of the natural substrate that have an excessive number of fluorescent dyes attached so that the fluorescence signal is almost non-existent. This quenching of the signal is caused by the close proximity of the dyes on the intact substrate. The enzyme-driven hydrolysis of the substrate results in separation of the dye results in separation of the dye molecules from one another and the fluorescence signal increases.

See User Manual for solubility instructions.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store in freezer (-5°C to -30°C) and protect from light.
Lyophilized
1 mg
Protein-Based Substrate
Room Temperature
Fluorescent
DQ™
Protease Substrate
Metalloproteinase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.