Learn More
Invitrogen™ DQ™ Collagen, type IV From Human Placenta, Fluorescein Conjugate
Analog of the natural substrate
Brand: Invitrogen™ D12052
Code : 50
Additional Details : Weight : 0.00525kg
Description
The fluorogenic DQ™ collagen can be used to directly monitor collagenase activity. DQ™ substrates are analogs of the natural substrate that have an excessive number of fluorescent dyes attached so that the fluorescence signal is almost non-existent. This quenching of the signal is caused by the close proximity of the dyes on the intact substrate. The enzyme-driven hydrolysis of the substrate results in separation of the dye results in separation of the dye molecules from one another and the fluorescence signal increases.
Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Chemotaxis and Cell Migration, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Lyophilized | |
1 mg | |
Protein-Based Substrate | |
Room Temperature |
Fluorescent | |
DQ™ | |
Protease Substrate | |
Metalloproteinase |
For Research Use Only. Not for use in diagnostic procedures.