Learn More
Invitrogen™ DQ™ Ovalbumin - Special Packaging
Description
DQ ovalbumin is a fluorogenic substrate for proteases. A strong fluorescence quenching effect is observed when proteins are heavily labeled with BODIPY dyes. Upon hydrolysis of the DQ ovalbumin to single, dye-labeled peptides by proteases, this quenching is relieved, producing brightly fluorescent products.
Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| Form | Solid |
| Product Line | DQ |
| Quantity | 5 x 1 mg |
| Substrate | Protease Substrate |
| Substrate Properties | Protein-Based Substrate |
| Target Enzyme | Proteases |
| Label or Dye | BODIPY™ FL |
| Product Type | Antigen processing and presentation studies |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.