missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EnzChek™ Lipase Substrate, green fluorescent, 505/515
Green-fluorescent product
Brand: Invitrogen™ E33955
This item is not returnable.
View return policy
Description
The nonfluorescent EnzChek™ lipase substrate, in the presence of lipases, produces a green-fluorescent product (excitation/emission maxima ∼507/515) for accurate lipase detection in solution.
Dissolve in high-quality anhydrous DMSO or chloroform to prepare a stock concentrations of up to 1 mM.
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5°C to -30°C) and protect from light. | |
Solid | |
100 μg | |
Chemical Substrate | |
1011.15 g/mol |
Fluorescence | |
EnzChek™ | |
Lipase substrate | |
507/515 nm | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.