Learn More
Invitrogen™ Epidermal Growth Factor, Biotin-XX Conjugate (biotin EGF)
This purified epidermal growth factor (EGF) contains a single biotin-XX molecule on the N-terminal amino acid
Brand: Invitrogen™ E3477
Code : 50
Additional Details : Weight : 0.00525kg
Description
- Biotin-XX EGF contains a long spacer arm that enhances the probe affinity for the EGF receptor and facilitates binding of a label such as a dye-streptavidin molecule, a Qdot™ nanocrystals-streptavidin molecule, or an enzyme-conjugated streptavidin
- Labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more
- EGF molecule: 53 amino acids, MW=6,045 Da
- Number of biotin-XX molecules on each EGF molecule: 1
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Wet Ice
Specifications
Store in freezer (-5 to -30°C). | |
Solid | |
Biotin & Other Haptens |
Fluorescence | |
Growth Factor | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.