Learn More
Invitrogen™ Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 555 Streptavidin (Alexa Fluor™ 555 EGF complex)
Complexes of the dye-streptavidin molecule and biotinylated EGF
Brand: Invitrogen™ E35350
Code : 50
Additional Details : Weight : 0.00525kg
Description
Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.
- Biotinylated conjugates utilize biotin-XX, which contains a long spacer arm designed to enhance the probe affinity for the EGF receptor
- EGF molecule: 53 amino acids, MW=6,045 Da
- Number of fluorophore molecules on each EGF molecule: 1
- Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Wet Ice
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Fluorescence | |
Growth Factor | |
Alexa Fluor™ |
Mammalian Cells | |
Lyophilized | |
Alexa Fluor Dyes | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.