missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epidermal Growth Factor, Biotinylated, complexed to Texas Red™ Streptavidin (Texas Red™ EGF complex)
Complexes of the dye-streptavidin molecule and biotinylated EGF
Brand: Invitrogen™ E3480
322.74 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.
- Biotinylated conjugates utilize biotin-XX, which contains long spacer arm designed to enhance probe affinity for EGF receptor
- EGF molecule: 53 amino acids, MW=6,045 Da
- Number of fluorophore molecules on each EGF molecule: 1
- Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter
Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Wet Ice
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Fluorescence | |
Classic Dyes | |
100 μg |
Mammalian Cells | |
Growth Factor | |
Texas Red™ | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.