Learn More
Invitrogen™ Molecular Probes™ Epidermal Growth Factor (EGF) (from Mouse Submaxillary Glands)
Epidermal growth factor (EGF) is a 53 amino acid (6,045 Da) protein
Brand: Invitrogen™ E3476
Code : 50
Additional Details : Weight : 0.00525kg
Description
- Unlabeled version may be used as competitor molecule (control) in experiments that use labeled EGF to study receptor binding
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Wet Ice