Learn More
Invitrogen™ Epidermal Growth Factor, Fluorescein Conjugate (Fluorescein EGF)
Constructed with the dye directly attached to EGF
Brand: Invitrogen™ E3478
Code : 50
Additional Details : Weight : 0.00525kg
Description
Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.
- EGF molecule: 53 amino acids, MW=6,045 Da
- Label (Ex/Em): Fluorescein (approx. 494/518nm)
- Number of fluorophore molecules on each EGF molecule: 1
- Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter
Order Info
Shipping Condition: Wet Ice
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Solid | |
Classic Dyes |
Fluorescence | |
Growth Factor | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.