missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibrinogen From Human Plasma, Alexa Fluor™ 594 Conjugate

For investigating platelet activation and subsequent fibrinogen binding

Brand:  Invitrogen™ F13193

236.36 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 11514776

  • £311.00 / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price

This item is not returnable. View return policy

Description

Description

Molecular Probes fibrinogen conjugates are prepared by attaching fluorescent dye molecules to purified human fibrinogen (approximately 15 dye molecules for each fibrinogen molecule), purifying the conjugate to remove unreacted dye, and then lyophilizing for storage. Fluorescently labeled fibrinogen has proven to be a valuable tool for investigating platelet activation and subsequent fibrinogen binding. For instance, fluorescein-labeled fibrinogen has been used to detect fibrinogen bound to activated platelets by flow cytometry.

  • Label (Ex/Em): Alexa Fluor 594 (approx. 590/617nm)
  • Spectrally similar to Texas Red™ dye conjugates, but brighter
  • Lyophilized product can be dissolved in buffer (e.g., sodium bicarbonate, pH 8.3) for use
  • Fluorescence is typically detected using fluorescence microscopy or flow cytometry

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mg
Human
Alexa Fluor Dyes
Lyophilized
Alexa Fluor™
Alexa Fluor 594
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
Fibrinogen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.