Learn More
Invitrogen™ FluoCells™ Prepared Slide #3 (mouse kidney section with Alexa Fluor™ 488 WGA, Alexa Fluor™ 568 Phalloidin, and DAPI)
Multilabeled cell preparations for observation by fluorescence microscopy validated for immunofluorescence
Brand: Invitrogen™ F24630
253.08 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
259.16 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
FluoCells prepared slide #3 is ideal for demonstrating the optical sectioning properties of confocal microscopes. Each slide contains a 16 μm cryostat section of mouse kidney stained with Alexa Fluor 488 wheat germ agglutinin (W-11261), Alexa Fluor 568 phalloidin (A-12380) and DAPI (D-1306).
Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)
Order Info
Shipping Condition: Room temperature

Specifications
Prepared Microscope Slide | |
Immunofluorescence | |
1 Each |
Alexa Fluor FluoCells | |
Store in freezer -5°C to -30°C and protect from light. | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.