Learn More
Invitrogen™ Fluorescein DHPE (N-(Fluorescein-5-Thiocarbamoyl)-1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)
Excitation/emission maxima ∽496/519nm
Brand: Invitrogen™ F362
154.67 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00525kg
Description
The phospholipid, fluorescein DHPE is labeled on the head group with the green-fluorescent pH-sensitive fluorophore, fluorescein
Cell Analysis, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Ionic Homeostasis & Signaling, pH Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Phospholipids | |
5 mg | |
Solid | |
Invitrogen |
For Research Use Only. Not for use in diagnostic procedures.