missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein - NIST-Traceable Standard - Nominal concentration 50μM - Special Packaging

Suitable for use in enzyme, protein activity assays and labeling chemistry

Brand:  Invitrogen™ F36915

318.44 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 11534826

  • £419.00 / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The NIST-traceable fluorescein standard is supplied as a calibrated 50μM solution of fluorescein in 100 mM sodium borate buffer, pH 9.5. Under these conditions, fluorescein is completely ionized and is therefore in its most fluorescent form. This standard meets the stringent criteria established by the National Institute of Standards and Technology (NIST) and is directly traceable to Standard Reference Material 1932 (SRM 1932). The NIST-traceable fluorescent standard can be used to assess day-to-day or experiment-to-experiment variation in fluorescence-based instrumentation as well as to determine the Molecules of Equivalent Soluble Fluorophore (MESF) value for an experimental solution.

  • Label type: Classsic dyes

Cell Analysis, Enzyme and Protein Activity Assays, Labeling Chemistry, Labeling QC and Calibration, Protein Assay Controls, Reference Standards and Accessories, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fluorescein Traceable Standard
Store in refrigerator 2°C to 8°C and protect from light.
Room Temperature
Classic Dyes
5 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.