Learn More
Invitrogen™ Fluorescein - Reference Standard
For use in cell viability, proliferation and function, enzyme and protein activity assays and labeling chemistry
Brand: Invitrogen™ F1300
94.67 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Fluorescein can be used as a reference standard for ether, ester and phosphate substrates derived from this green-fluorescent dye.
- Label type: Classsic dyes
Cell Analysis, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Ionic Homeostasis and Signaling, Labeling Chemistry, Labeling QC and Calibration, Protein Assay Controls, Reference Standards and Accessories, Proteins, Expression, Isolation and Analysis, pH Detection
Order Info
Shipping Condition: Room temperature
Specifications
Fluorescein Reference Standard | |
Classic Dyes | |
1 g |
Green | |
Store at room temperature and protect from light. | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.